1,1,3,3-Tetramethylbutylisocyanid 95%

Lieferant: Apollo Scientific

Synonyme: 1,1,3,3-Tetramethylbutylisocyanide

OR927941-25G OR927941-5G
APOSOR927941-25GEA 802 EUR
APOSOR927941-25G APOSOR927941-5G
1,1,3,3-Tetramethylbutylisocyanid 95%
1,1,3,3-Tetramethylbutylisocyanid
Formel: C₉H₁₇N
Molecular Weight: 139,2380 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00000003
CAS-Nummer: 14542-93-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR