(3-Propoxyphenyl)methanamine hydrochloride 95%
Lieferant: Apollo Scientific
Synonyme:
(3-Propoxyphenyl)methanamine
OR927946-1G
OR927946-250MG
APOSOR927946-1GEA
415
EUR
APOSOR927946-1G
APOSOR927946-250MG
(3-Propoxyphenyl)methanamine hydrochloride 95%
(3-Propoxyphenyl)methanamine hydrochloride
Formel:
C₁₀H₁₆ClNO Molecular Weight: 201,7 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10687270 CAS-Nummer: 37806-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...