(E)-3-Methoxy-2-butenoic acid methyl ester 95%
Lieferant: Apollo Scientific
OR927974-1G OR927974-250MG
APOSOR927974-1GEA 574 EUR
APOSOR927974-1G APOSOR927974-250MG
(E)-3-Methoxy-2-butenoic acid methyl ester 95%
(E)-3-Methoxy-2-butenoic acid methyl ester
Formel: C₆H₁₀O₃
Molecular Weight: 130.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09838566
CAS-Nummer: 4525-28-4