1-Allyl-1H-pyrazole-4-carbaldehyde 95%

Lieferant: Apollo Scientific

OR928029-1G OR928029-250MG
APOSOR928029-1GEA 400 EUR
APOSOR928029-1G APOSOR928029-250MG
1-Allyl-1H-pyrazole-4-carbaldehyde 95%
1-Allyl-1H-pyrazole-4-carbaldehyde
Formel: C₇H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD12827481
CAS-Nummer: 78758-36-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR