1,3-Dimethyl-1H-indazol-5-amine 95%

Lieferant: Apollo Scientific

OR928039-1G OR928039-250MG
APOSOR928039-1GEA 133 EUR
APOSOR928039-1G APOSOR928039-250MG
1,3-Dimethyl-1H-indazol-5-amine 95%
1,3-Dimethyl-1H-indazol-5-amine
Formel: C₉H₁₁N₃
Molecular Weight: 161,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20705804
CAS-Nummer: 5757-85-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR