1H-Imidazole-2-carbonitrile hydrochloride 95%

Lieferant: Apollo Scientific

OR928305-1G OR928305-250MG
APOSOR928305-1GEA 696 EUR
APOSOR928305-1G APOSOR928305-250MG
1H-Imidazole-2-carbonitrile hydrochloride 95%
1H-Imidazole-2-carbonitrile hydrochloride
Formel: C₄H₃N₃·HCl
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1392803-62-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR