4,4-Dimethoxybutannitril 95%
Lieferant: Apollo Scientific
Synonyme:
3-Cyanopropionaldehyde dimethyl Acetal, 4,4-Dimethoxybutyronitrile
OR928315-25G
OR928315-100G
APOSOR928315-25GEA
126
EUR
APOSOR928315-25G
APOSOR928315-100G
4,4-Dimethoxybutannitril 95%
4,4-Dimethoxybutanenitrile
Formel:
C₆H₁₁NO₂ Molecular Weight: 129.1570 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00019883 CAS-Nummer: 14618-78-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...