(1H-Benzoimidazol-2-ylmethyl)methylamine 98%
Lieferant: Apollo Scientific
Synonyme:
1-(1H-Benzo[d]imidazol-2-yl)-N-methylmethanamine
OR928418-1G
OR928418-250MG
OR928418-5G
APOSOR928418-1GEA
132
EUR
APOSOR928418-1G
APOSOR928418-250MG
APOSOR928418-5G
(1H-Benzoimidazol-2-ylmethyl)methylamine 98%
(1H-Benzoimidazol-2-ylmethyl)methylamine
Formel:
C₉H₁₁N₃ Molecular Weight: 161.21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06738854 CAS-Nummer: 98997-01-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...