1-Pyridazin-3-ylethanone
Lieferant: Apollo Scientific
OR928493-1G
OR928493-5G
APOSOR928493-1GEA
410
EUR
APOSOR928493-1G
APOSOR928493-5G
1-Pyridazin-3-ylethanone
1-Pyridazin-3-ylethanone
Formel:
C₆H₆N₂O Molecular Weight: 122,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13189577 CAS-Nummer: 1122-63-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...