L-Argininamide dihydrochloride 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: H-Arg-NH₂·2HCl, H-Arg-NH₂·2 HCl, H-Arg-NH2.2HCl

OR928586-1G OR928586-25G OR928586-5G
APOSOR928586-1GEA 68.3 EUR
APOSOR928586-1G APOSOR928586-25G APOSOR928586-5G
L-Argininamide dihydrochloride 97%
L-Argininamide dihydrochloride
Formel: C₆H₁₇Cl₂N₅O
Molecular Weight: 246,136 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00058286
CAS-Nummer: 14975-30-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR