Ethyl-4-ethoxybenzoat 98%

Lieferant: Apollo Scientific

Synonyme: 4-Ethoxybenzoesäureethylester

OR928615-100G
APOSOR928615-100GEA 156 EUR
APOSOR928615-100G
Ethyl-4-ethoxybenzoat 98%
Ethyl-4-ethoxybenzoat
Formel: C₁₁H₁₄O₃
Molecular Weight: 194,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009116
CAS-Nummer: 23676-09-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR