2-Ethylsulphanylpyrimidine-5-carbaldehyde 95%
Lieferant: Apollo Scientific
Synonyme:
2-(Ethylthio)pyrimidine-5-carbaldehyde
OR928707-1G
APOSOR928707-1GEA
200
EUR
APOSOR928707-1G
2-Ethylsulphanylpyrimidine-5-carbaldehyde 95%
2-Ethylsulphanylpyrimidine-5-carbaldehyde
Formel:
C₇H₈N₂OS Molecular Weight: 168,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07186316 CAS-Nummer: 876890-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...