2H-Indazol-2-ylacetic acid 95%

Lieferant: Apollo Scientific

OR928715-1G OR928715-250MG
APOSOR928715-1GEA 538 EUR
APOSOR928715-1G APOSOR928715-250MG
2H-Indazol-2-ylacetic acid 95%
2H-Indazol-2-ylacetic acid
Formel: C₉H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06739007
CAS-Nummer: 58037-05-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR