1-Allyl-3,5-dimethyl-1H-pyrazole-4-carbaldehyde 95%
Lieferant: Apollo Scientific
OR928727-1G
OR928727-5G
APOSOR928727-1GEA
168
EUR
APOSOR928727-1G
APOSOR928727-5G
1-Allyl-3,5-dimethyl-1H-pyrazole-4-carbaldehyde 95%
1-Allyl-3,5-dimethyl-1H-pyrazole-4-carbaldehyde
Formel:
C₉H₁₂N₂O Molecular Weight: 164.21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12103568 CAS-Nummer: 1155595-91-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...