6-Acetyl-2H-1,4-benzoxazin-3(4H)-on 95%
Lieferant: Apollo Scientific
Synonyme:
6-Acetyl-2H-benzo[b][1,4]oxazin-3(4H)-one
OR928736-25G
APOSOR928736-25GEA
173
EUR
APOSOR928736-25G
6-Acetyl-2H-1,4-benzoxazin-3(4H)-on 95%
6-Acetyl-2H-1,4-benzoxazin-3(4H)-on
Formel:
C₁₀H₉NO₃ Molecular Weight: 191,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01321313 CAS-Nummer: 26518-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...