Bis(dimethylamino)dimethylsilan 95%

Lieferant: Apollo Scientific

OR928817-100G OR928817-25G
APOSOR928817-100GEA 435 EUR
APOSOR928817-100G APOSOR928817-25G
Bis(dimethylamino)dimethylsilan 95%
Bis(dimethylamino)dimethylsilan
Formel: C₆H₁₈N₂Si
Molecular Weight: 146,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008296
CAS-Nummer: 3768-58-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR