(3,5-Dichloro-4-pyridyl)methanol 98%
Lieferant: Apollo Scientific
Synonyme:
(3,5-Dichloropyridin-4-yl)methanol
OR928943-1G
OR928943-250MG
APOSOR928943-1GEA
458
EUR
APOSOR928943-1G
APOSOR928943-250MG
(3,5-Dichloro-4-pyridyl)methanol 98%
(3,5-Dichloro-4-pyridyl)methanol
Formel:
C₆H₅Cl₂NO Molecular Weight: 178,02 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06202840 CAS-Nummer: 159783-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...