1-Ethyl-1H-indole-6-carbaldehyde 95%
Lieferant: Apollo Scientific
OR929089-1G
OR929089-5G
APOSOR929089-1GEA
226
EUR
APOSOR929089-1G
APOSOR929089-5G
1-Ethyl-1H-indole-6-carbaldehyde 95%
1-Ethyl-1H-indole-6-carbaldehyde
Formel:
C₁₁H₁₁NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD09455258 CAS-Nummer: 854778-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...