2-Acetyl-4-chlorthiophen 95%
Lieferant: Apollo Scientific
Synonyme:
1-(4-Chlor-2-thienyl)-1-ethanon
OR929222-1G
OR929222-5G
APOSOR929222-1GEA
26.1
EUR
APOSOR929222-1G
APOSOR929222-5G
2-Acetyl-4-chlorthiophen 95%
2-Acetyl-4-chlorthiophen
Formel:
C₆H₅ClOS Molecular Weight: 160,62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082791 CAS-Nummer: 34730-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...