1-Ethyl-1H-indole-3-carbaldehyde 95%

Lieferant: Apollo Scientific

OR929292-1G OR929292-5G
APOSOR929292-1GEA 78.3 EUR
APOSOR929292-1G APOSOR929292-5G
1-Ethyl-1H-indole-3-carbaldehyde 95%
1-Ethyl-1H-indole-3-carbaldehyde
Formel: C₁₁H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00047263
CAS-Nummer: 58494-59-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR