Tri-O-benzyl-D-glucal 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,4,6-Tri-O-benzyl-D-glucal, 1,5-Anhydro-2-deoxy-D-arabino-hex-1-enitol 3,4,6-Tribenzyl Ether
OR929303-25G
OR929303-5G
APOSOR929303-25GEA
519
EUR
APOSOR929303-25G
APOSOR929303-5G
Tri-O-benzyl-D-glucal 98%
Tri-O-benzyl-D-glucal
Formel:
C₂₇H₂₈O₄ Molecular Weight: 416.52 g/mol Schmelzpunkt: 55…59 °C Flammpunkt: >113 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00061640 CAS-Nummer: 55628-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...