2,3,4,6-Tetra-O-acetyl-D-glucopyranose 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,3,4,6-tetra-O-acetylglucopyranose, D-Glucopyranose 2,3,4,6-tetraacetate, (2R,3R,4S,5R)-2-(Acetoxymethyl)-6-hydroxytetrahydro-2H-pyran-3,4,5-triyl triacetate, 2,3,4,6-tetra-O-acetyl-beta-D-glucopyranose
OR929363-25G
OR929363-5G
APOSOR929363-25GEA
340
EUR
APOSOR929363-25G
APOSOR929363-5G
2,3,4,6-Tetra-O-acetyl-D-glucopyranose 96%
2,3,4,6-Tetra-O-acetyl-D-glucopyranose
Formel:
C₁₄H₂₀O₁₀ Molecular Weight: 348,304 g/mol Schmelzpunkt: 137…138 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00061634 CAS-Nummer: 10343-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...