p,α-Dimethylbenzylalkohol 95%

Lieferant: Apollo Scientific

Synonyme: 1-(p-Tolyl)ethanol, 1-(4-Methylphenyl)ethanol, α,4-Dimethylbenzylalkohol, 4-Methylphenyl-methylcarbinol, 4-(1-Hydroxyethyl)toluol

OR929381-100G OR929381-25G
APOSOR929381-100GEA 177 EUR
APOSOR929381-100G APOSOR929381-25G
p,α-Dimethylbenzylalkohol 95%
p,α-Dimethylbenzylalkohol
Formel: C₉H₁₂O
Molecular Weight: 136,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016860
CAS-Nummer: 536-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR