Methyl pyridazine-3-carboxylate 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR929425-1G
OR929425-5G
APOSOR929425-1GEA
56.7
EUR
APOSOR929425-1G
APOSOR929425-5G
Methyl pyridazine-3-carboxylate 95%
Methyl pyridazine-3-carboxylate
Formel:
C₆H₆N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD10698044 CAS-Nummer: 34253-02-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...