4-Methyl-2H-1,4-benzoxazin-3(4H)-one 95%

Lieferant: Apollo Scientific

Synonyme: 4-Methyl-2H-1,4-Benzoxazin-3-one

OR929548-25G
APOSOR929548-25GEA 114 EUR
APOSOR929548-25G
4-Methyl-2H-1,4-benzoxazin-3(4H)-one 95%
4-Methyl-2H-1,4-benzoxazin-3(4H)-one
Formel: C₉H₉NO₂
Molecular Weight: 163,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00601365
CAS-Nummer: 21744-84-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR