4-(3-Hydroxy-3-methylbut-1-ynyl)benzaldehyde 95%
Lieferant: Apollo Scientific
OR929616-1G
OR929616-5G
APOSOR929616-1GEA
168
EUR
APOSOR929616-1G
APOSOR929616-5G
4-(3-Hydroxy-3-methylbut-1-ynyl)benzaldehyde 95%
4-(3-Hydroxy-3-methylbut-1-ynyl)benzaldehyde
Formel:
C₁₂H₁₂O₂ Molecular Weight: 188,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07187866 CAS-Nummer: 117569-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...