4-Phenylpyridin-2-amine 98%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-4-phenylpyridine

OR929662-1G OR929662-5G
APOSOR929662-1GEA 134 EUR
APOSOR929662-1G APOSOR929662-5G
4-Phenylpyridin-2-amine 98%
4-Phenylpyridin-2-amine
Formel: C₁₁H₁₀N₂
Molecular Weight: 170,2105 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00137848
CAS-Nummer: 60781-83-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR