3-Chlor-5-methyl-4-hydroxybenzonitril 98%

Lieferant: Apollo Scientific

OR929667-10G OR929667-1G OR929667-5G
APOSOR929667-10GEA 451 EUR
APOSOR929667-10G APOSOR929667-1G APOSOR929667-5G
3-Chlor-5-methyl-4-hydroxybenzonitril 98%
3-Chlor-5-methyl-4-hydroxybenzonitril
Formel: C₈H₆ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD08689583
CAS-Nummer: 173900-45-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR