[(1-Isopropylpiperidin-4-yl)methyl]amine 96%
Lieferant: Apollo Scientific
OR929697-1G
OR929697-250MG
APOSOR929697-1GEA
377
EUR
APOSOR929697-1G
APOSOR929697-250MG
[(1-Isopropylpiperidin-4-yl)methyl]amine 96%
[(1-Isopropylpiperidin-4-yl)methyl]amine
Formel:
C₉H₂₀N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07365293 CAS-Nummer: 132740-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...