2-Methyl-5,6-dihydro-4H-cyclopenta[d][1,3]thiazole-4-carboxylic acid 95%

Lieferant: Apollo Scientific

Synonyme: 2-Methyl-4h,5h,6h-cyclopenta[d][1,3]thiazole-4-carboxylic acid

OR929733-100MG OR929733-1G OR929733-250MG
APOSOR929733-100MGEA 161 EUR
APOSOR929733-100MG APOSOR929733-1G APOSOR929733-250MG
2-Methyl-5,6-dihydro-4H-cyclopenta[d][1,3]thiazole-4-carboxylic acid 95%
2-Methyl-5,6-dihydro-4H-cyclopenta[d][1,3]thiazole-4-carboxylic acid
Formel: C₈H₉NO₂S
Molecular Weight: 183,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19706035
CAS-Nummer: 1190391-79-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR