2-Acetyl-1-ethylpyrrole 98%
Lieferant: Apollo Scientific
Synonyme:
1-(1-Ethyl-1H-pyrrol-2-yl)ethanone
OR929782-100G
OR929782-25G
APOSOR929782-100GEA
179
EUR
APOSOR929782-100G
APOSOR929782-25G
2-Acetyl-1-ethylpyrrole 98%
2-Acetyl-1-ethylpyrrole
Formel:
C₈H₁₁NO Molecular Weight: 137,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191234 CAS-Nummer: 39741-41-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...