5-Chloro-2-methylpyrimidine 95%
Lieferant: Apollo Scientific
OR929840-1G
OR929840-250MG
APOSOR929840-1GEA
130
EUR
APOSOR929840-1G
APOSOR929840-250MG
5-Chloro-2-methylpyrimidine 95%
5-Chloro-2-methylpyrimidine
Formel:
C5H5ClN2 Molecular Weight: 128,5596 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10574686 CAS-Nummer: 54198-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...