4-Formyl-3-hydroxybenzoesäure 95%
Lieferant: Apollo Scientific
OR929939-1G
OR929939-5G
APOSOR929939-1GEA
134
EUR
APOSOR929939-1G
APOSOR929939-5G
4-Formyl-3-hydroxybenzoesäure 95%
4-Formyl-3-hydroxybenzoic acid
Formel:
C₈H₆O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD05865188 CAS-Nummer: 619-12-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...