2-Chlor-α-methylbenzylalkohol 95%

Lieferant: Apollo Scientific

Synonyme: 1-(2-Chlorophenyl)ethanol, o-Chloro-α-methylbenzyl alcohol, 2-Chlorophenyl methyl carbinol

OR929968-100G OR929968-25G
APOSOR929968-100GEA 362 EUR
APOSOR929968-100G APOSOR929968-25G
2-Chlor-α-methylbenzylalkohol 95%
2-Chlor-α-methylbenzylalkohol
Formel: C₈H₉ClO
Molecular Weight: 156,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00041037
CAS-Nummer: 13524-04-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR