4-[5-(Chloromethyl)-1,2,4-oxadiazol-3-yl]pyridine 95%

Lieferant: Apollo Scientific

OR929999-1G OR929999-250MG
APOSOR929999-1GEA 377 EUR
APOSOR929999-1G APOSOR929999-250MG
4-[5-(Chloromethyl)-1,2,4-oxadiazol-3-yl]pyridine 95%
4-[5-(Chloromethyl)-1,2,4-oxadiazol-3-yl]pyridine
Formel: C₈H₆ClN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD09034189
CAS-Nummer: 50737-35-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR