N-Methyl-7H-purin-6-amine 95%

Lieferant: Apollo Scientific

Synonyme: N6-Methyladenine, 6-Methylaminopurine

OR930003-10G OR930003-1G OR930003-5G
APOSOR930003-10GEA 752 EUR
APOSOR930003-10G APOSOR930003-1G APOSOR930003-5G
N-Methyl-7H-purin-6-amine 95%
N-Methyl-7H-purin-6-amine
Formel: C₆H₇N₅
Molecular Weight: 149,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00075820
CAS-Nummer: 443-72-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR