4-Pyridazinemethanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Pyridazin-4-ylmethanol
OR930101-1G
OR930101-250MG
APOSOR930101-1GEA
394
EUR
APOSOR930101-1G
APOSOR930101-250MG
4-Pyridazinemethanol 97%
4-Pyridazinemethanol
Formel:
C₅H₆N₂O Molecular Weight: 110,12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06227446 CAS-Nummer: 50901-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...