2-tert-Butoxybenzaldehyd 95%
Lieferant: Apollo Scientific
OR930137-1G
OR930137-250MG
APOSOR930137-1GEA
647
EUR
APOSOR930137-1G
APOSOR930137-250MG
2-tert-Butoxybenzaldehyd 95%
2-tert-Butoxybenzaldehyd
Formel:
C₁₁H₁₄O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD02258844 CAS-Nummer: 35129-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...