N,N-Dimethyl(2-methyl-1H-pyrrolo[2,3-b]pyridin-3-yl)methanamine 95%

Lieferant: Apollo Scientific

OR930160-1G OR930160-250MG
APOSOR930160-1GEA 647 EUR
APOSOR930160-1G APOSOR930160-250MG
N,N-Dimethyl(2-methyl-1H-pyrrolo[2,3-b]pyridin-3-yl)methanamine 95%
N,N-Dimethyl(2-methyl-1H-pyrrolo[2,3-b]pyridin-3-yl)methanamine
Formel: C₁₁H₁₅N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD13191648
CAS-Nummer: 7546-48-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR