Ethyl-5,6-dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylate 95%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 5,6-dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylate
OR930225-1G
APOSOR930225-1GEA
62.5
EUR
APOSOR930225-1G
Ethyl-5,6-dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylate 95%
Ethyl-5,6-dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylate
Formel:
C₉H₁₂N₂O₂ Molecular Weight: 180,2000 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11975789 CAS-Nummer: 86477-09-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...