tert-Butyldimethyl(tributylstannylmethoxy)silane 95%

Lieferant: Apollo Scientific

OR930278-1G OR930278-250MG OR930278-5G
APOSOR930278-1GEA 208 EUR
APOSOR930278-1G APOSOR930278-250MG APOSOR930278-5G
tert-Butyldimethyl(tributylstannylmethoxy)silane 95%
tert-Butyldimethyl(tributylstannylmethoxy)silane
Formel: C₁₉H₄₄OSiSn
Molecular Weight: 435,34 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD23152698
CAS-Nummer: 123061-64-3
UN: 2788
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR