2,2-Diethoxypropan 95%
Lieferant: Apollo Scientific
Synonyme:
Acetondiethylacetal
OR930301-25G
OR930301-5G
OR930301-100G
APOSOR930301-25GEA
270
EUR
APOSOR930301-25G
APOSOR930301-5G
APOSOR930301-100G
2,2-Diethoxypropan 95%
2,2-Diethoxypropane
Formel:
C₇H₁₆O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009224 CAS-Nummer: 126-84-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...