2-(Ethoxymethyl)furan 95%
Lieferant: Apollo Scientific
OR930324-10G
OR930324-25G
APOSOR930324-10GEA
133
EUR
APOSOR930324-10G
APOSOR930324-25G
2-(Ethoxymethyl)furan 95%
2-(Ethoxymethyl)furan
Formel:
C₇H₁₀O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD02258625 CAS-Nummer: 6270-56-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...