2-Chlor-3'-methoxyacetanilid 95%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-N-(3-methoxyphenyl)acetamide

OR930471-1G OR930471-5G
APOSOR930471-1GEA 78.3 EUR
APOSOR930471-1G APOSOR930471-5G
2-Chlor-3'-methoxyacetanilid 95%
2-Chlor-3'-methoxyacetanilid
Formel: C₉H₁₀ClNO₂
Molecular Weight: 199,6342 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00018900
CAS-Nummer: 17641-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR