3-Amino-2-isopropyl-5-methyl-2H-pyrazole 95%
Lieferant: Apollo Scientific
Synonyme:
5-Amino-1-isopropyl-3-methylpyrazole, 1-Isopropyl-3-methyl-1H-pyrazol-5-amine
OR930472-1G
OR930472-5G
APOSOR930472-1GEA
101
EUR
APOSOR930472-1G
APOSOR930472-5G
3-Amino-2-isopropyl-5-methyl-2H-pyrazole 95%
3-Amino-2-isopropyl-5-methyl-2H-pyrazole
Formel:
C₇H₁₃N₃ Molecular Weight: 139.202 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00514520 CAS-Nummer: 1124-16-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...