(4-Formylphenoxy)essigsäure 98%
Lieferant: Apollo Scientific
Synonyme:
2-(4-Formylphenoxy)acetic acid
OR930525-100G
OR930525-25G
APOSOR930525-100GEA
374
EUR
APOSOR930525-100G
APOSOR930525-25G
(4-Formylphenoxy)essigsäure 98%
(4-Formylphenoxy)essigsäure
Formel:
(C₆H₄(CHO))OCH₂COOH Molecular Weight: 180,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016613 CAS-Nummer: 22042-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...