5-Chloro-1H-indole-2-carbaldehyde 95%

Lieferant: Apollo Scientific

OR930619-1G OR930619-250MG
APOSOR930619-1GEA 154 EUR
APOSOR930619-1G APOSOR930619-250MG
5-Chloro-1H-indole-2-carbaldehyde 95%
5-Chloro-1H-indole-2-carbaldehyde
Formel: C₉H₆ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD06800428
CAS-Nummer: 53590-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR