3-(Methylthio)benzaldehyd 95%
Lieferant: Apollo Scientific
OR930751-1G
OR930751-250MG
APOSOR930751-1GEA
159
EUR
APOSOR930751-1G
APOSOR930751-250MG
3-(Methylthio)benzaldehyd 95%
3-(Methylthio)benzaldehyde
Formel:
C₈H₈OS Molecular Weight: 152,213 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09864229 CAS-Nummer: 73771-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...