2,4-Diethoxybenzaldehyd 95%
Lieferant: Apollo Scientific
OR930819-5G
APOSOR930819-5GEA
59.6
EUR
APOSOR930819-5G
2,4-Diethoxybenzaldehyd 95%
2,4-Diethoxybenzaldehyde
Formel:
C₁₁H₁₄O₃ Molecular Weight: 194,2271 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00995010 CAS-Nummer: 22924-16-9 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...