4-(Pyrrolidin-1-ylcarbonyl)pyridin-2-amine 95%

Lieferant: Apollo Scientific

OR930925-1G OR930925-250MG OR930925-5G
APOSOR930925-1GEA 197 EUR
APOSOR930925-1G APOSOR930925-250MG APOSOR930925-5G
4-(Pyrrolidin-1-ylcarbonyl)pyridin-2-amine 95%
4-(Pyrrolidin-1-ylcarbonyl)pyridin-2-amine
Formel: C₁₀H₁₃N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD11053954
CAS-Nummer: 1060817-34-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR